3-Bromo-2-chloro-5-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

85067-500MG 85067-1G 85067-5G
77337-632EA 431.57 USD
77337-632 77358-958 77382-926
3-Bromo-2-chloro-5-(trifluoromethoxy)benzoic acid ≥97%
3-Bromo-2-chloro-5-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₃BrClF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34770339
CAS Number: 2168045-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR