2-((4-Bromophenyl)ethynyl)aniline ≥95%

Supplier: AOBChem USA

Warning

85031-500MG 85031-1G 85031-5G
77334-798EA 320.33 USD
77334-798 77358-954 77382-922
2-((4-Bromophenyl)ethynyl)aniline ≥95%
2-((4-Bromophenyl)ethynyl)aniline

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrN
MW: 272.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00168850
CAS Number: 157869-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR