2-((3-Bromophenyl)ethynyl)aniline ≥97%

Supplier: AOBChem USA

Warning

85029-500MG 85029-1G 85029-5G 85029-25G
77334-796EA 161.71 USD
77334-796 77346-028 77370-834 77391-422
2-((3-Bromophenyl)ethynyl)aniline ≥97%
2-((3-Bromophenyl)ethynyl)aniline

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrN
MW: 272.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23141116
CAS Number: 1438414-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR