1-Cyclohexyl-4-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

85021-500MG 85021-1G 85021-5G
77339-308EA 575.77 USD
77339-308 77358-952 77382-918
1-Cyclohexyl-4-fluorobenzene ≥97%
1-Cyclohexyl-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₅F
Storage Temperature: Ambient
MDL Number: MFCD15527405
CAS Number: 1717-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR