3,4-Dichlorophenyl acetate ≥97%

Supplier: AOBChem USA

Warning

85015-500MG 85015-1G 85015-5G
77334-790EA 348.14 USD
77334-790 77358-948 77382-914
3,4-Dichlorophenyl acetate ≥97%
3,4-Dichlorophenyl acetate

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD01632288
CAS Number: 17847-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR