4-Bromo-2-ethoxyphenol ≥95%

Supplier: AOBChem USA

Warning

84702-500MG 84702-1G 84702-5G 84702-25G
77334-786EA 145.23 USD
77334-786 77345-114 77370-826 77391-420
4-Bromo-2-ethoxyphenol ≥95%
4-Bromo-2-ethoxyphenol

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD09907051
CAS Number: 211495-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR