2-(Benzyloxy)-4-bromo-1-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

84700-1G 84700-5G 84700-25G
77340-838EA 60.77 USD
77340-838 77370-824 77382-906
2-(Benzyloxy)-4-bromo-1-iodobenzene ≥97%
2-(Benzyloxy)-4-bromo-1-iodobenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrIO
Storage Temperature: Ambient
MDL Number: MFCD34770127
CAS Number: 1632373-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR