4-Bromo-1-iodo-2-isobutoxybenzene ≥97%

Supplier: AOBChem USA

Warning

84699-1G 84699-5G 84699-25G
77340-922EA 62.83 USD
77340-922 77370-822 77382-904
4-Bromo-1-iodo-2-isobutoxybenzene ≥97%
4-Bromo-1-iodo-2-isobutoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrIO
Storage Temperature: Ambient
MDL Number: MFCD34770126
CAS Number: 2901103-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR