4-Bromo-1-iodo-2-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

84698-500MG 84698-1G 84698-5G 84698-25G
77334-784EA 161.71 USD
77334-784 77346-026 77370-820 77391-418
4-Bromo-1-iodo-2-isopropoxybenzene ≥97%
4-Bromo-1-iodo-2-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrIO
Storage Temperature: Ambient
MDL Number: MFCD12547879
CAS Number: 1208075-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR