4-Bromo-2-ethyl-6-methylbenzonitrile ≥95%

Supplier: AOBChem USA

Warning

84637-500MG 84637-1G 84637-5G
77336-986EA 402.73 USD
77336-986 77358-944 77382-902
4-Bromo-2-ethyl-6-methylbenzonitrile ≥95%
4-Bromo-2-ethyl-6-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrN
Storage Temperature: Ambient
MDL Number: MFCD18917120
CAS Number: 1349717-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR