1-Ethoxy-3,5-bis(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

84568-1G 84568-5G 84568-25G
77342-082EA 112.27 USD
77342-082 77370-816 77382-898
1-Ethoxy-3,5-bis(trifluoromethyl)benzene ≥97%
1-Ethoxy-3,5-bis(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₈F₆O
MW: 258.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28396334
CAS Number: 368-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR