5-Bromo-1-ethoxy-2-fluoro-3-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

84430-500MG 84430-1G 84430-5G 84430-25G
77334-782EA 163.77 USD
77334-782 77346-136 77370-814 77391-416
5-Bromo-1-ethoxy-2-fluoro-3-(methylsulfonyl)benzene ≥97%
5-Bromo-1-ethoxy-2-fluoro-3-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO₃S
Storage Temperature: Ambient
MDL Number: MFCD34770009
CAS Number: 2901102-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR