2,2-Dichloro-1-(2-fluoro-5-(trifluoromethyl)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

84202-500MG 84202-1G 84202-5G 84202-25G
77334-778EA 179.22 USD
77334-778 77346-748 77370-810 77391-412
2,2-Dichloro-1-(2-fluoro-5-(trifluoromethyl)phenyl)ethanone ≥97%
2,2-Dichloro-1-(2-fluoro-5-(trifluoromethyl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₄Cl₂F₄O
MW: 275.027 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769871
CAS Number: 2901099-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR