2,2-Dichloro-1-(5-chloro-2-(trifluoromethyl)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

84201-500MG 84201-1G 84201-5G 84201-25G
77334-776EA 101.97 USD
77334-776 77343-020 77370-808 77382-892
2,2-Dichloro-1-(5-chloro-2-(trifluoromethyl)phenyl)ethanone ≥97%
2,2-Dichloro-1-(5-chloro-2-(trifluoromethyl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₄Cl₃F₃O
MW: 291.482 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769870
CAS Number: 2901102-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR