6-Fluoro-2H-chromen-2-one ≥95%

Supplier: AOBChem USA

Warning

84186-500MG 84186-1G 84186-5G 84186-25G
77334-772EA 174.07 USD
77334-772 77346-466 77370-806 77391-408
6-Fluoro-2H-chromen-2-one ≥95%
6-Fluoro-2H-chromen-2-one

Precaución: for Research Use Only

Formula: C₉H₅FO₂
Storage Temperature: Ambient
MDL Number: MFCD08753855
CAS Number: 75487-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR