1-Bromo-2-chloro-5-ethoxy-4-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

83966-500MG 83966-1G 83966-5G 83966-25G
77334-770EA 280.16 USD
77334-770 77358-932 77370-802 77391-406
1-Bromo-2-chloro-5-ethoxy-4-fluorobenzene ≥97%
1-Bromo-2-chloro-5-ethoxy-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₇BrClFO
Storage Temperature: Ambient
MDL Number: MFCD34769745
CAS Number: 2387210-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR