Methyl 5-iodo-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

83939-500MG 83939-1G 83939-5G 83939-25G
77334-768EA 214.24 USD
77334-768 77358-930 77370-798 77391-404
Methyl 5-iodo-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥97%
Methyl 5-iodo-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₆F₃IO₂S
MW: 386.129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29399583
CAS Number: 2901066-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR