Methyl 7-bromo-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

83936-500MG 83936-1G 83936-25G 83936-5G
77334-766EA 291.49 USD
77334-766 77358-928 77391-402 77382-880
Methyl 7-bromo-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥97%
Methyl 7-bromo-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₆BrF₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD29663350
CAS Number: 2901103-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR