Methyl 3,7-bis(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

83894-500MG 83894-1G 83894-5G
77339-428EA 588.13 USD
77339-428 77358-926 77382-878
Methyl 3,7-bis(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥97%
Methyl 3,7-bis(trifluoromethyl)benzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₆F₆O₂S
MW: 328.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29691007
CAS Number: 2901085-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR