3-(5-Bromo-2-chloropyridin-4-yl)-1,1,1-trifluoropropan-2-ol ≥97%

Supplier: AOBChem USA

Warning

83885-500MG 83885-1G 83885-5G
77337-714EA 437.75 USD
77337-714 77358-924 77382-876
3-(5-Bromo-2-chloropyridin-4-yl)-1,1,1-trifluoropropan-2-ol ≥97%
3-(5-Bromo-2-chloropyridin-4-yl)-1,1,1-trifluoropropan-2-ol

Precaución: for Research Use Only

Formula: C₈H₆BrClF₃NO
MW: 304.492 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769705
CAS Number: 2353599-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR