1-(3,5-Dichlorophenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

83826-500MG 83826-1G 83826-5G 83826-25G
77334-764EA 116.39 USD
77334-764 77343-516 77370-796 77382-874
1-(3,5-Dichlorophenyl)cyclohexanol ≥95%
1-(3,5-Dichlorophenyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₂H₁₄Cl₂O
MW: 245.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769659
CAS Number: 77826-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR