1-(3,5-Dichlorophenyl)cyclopentanol ≥97%

Supplier: AOBChem USA

Warning

83825-500MG 83825-1G 83825-5G 83825-25G
77334-762EA 161.71 USD
77334-762 77346-024 77370-794 77391-400
1-(3,5-Dichlorophenyl)cyclopentanol ≥97%
1-(3,5-Dichlorophenyl)cyclopentanol

Precaución: for Research Use Only

Formula: C₁₁H₁₂Cl₂O
MW: 231.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769658
CAS Number: 1522942-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR