3-(phenylethynyl)pyridin-2-amine ≥97%

Supplier: AOBChem USA

Warning

83798-500MG 83798-1G 83798-5G 83798-25G
77334-758EA 169.95 USD
77334-758 77346-384 77370-792 77391-396
3-(phenylethynyl)pyridin-2-amine ≥97%
3-(phenylethynyl)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₀N₂
Storage Temperature: Ambient
MDL Number: MFCD20721090
CAS Number: 950917-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR