(2-Chloro-6-iodophenyl)(methyl)sulfane ≥97%
Supplier: AOBChem USA
|
Warning
|
83741-500MG
83741-1G
77340-556EA
1818.98
USD
77340-556
77358-918
(2-Chloro-6-iodophenyl)(methyl)sulfane ≥97%
(2-Chloro-6-iodophenyl)(methyl)sulfane
Precaución: for Research Use Only
Formula:
C₇H₆ClIS Storage Temperature: Ambient |
MDL Number:
MFCD22489411 CAS Number: 1822764-11-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...