2-(Benzyloxy)-3-ethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

83452-500MG 83452-1G 83452-5G 83452-25G
77334-748EA 202.91 USD
77334-748 77358-914 77370-782 77391-390
2-(Benzyloxy)-3-ethylbenzoic acid ≥97%
2-(Benzyloxy)-3-ethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD34769527
CAS Number: 2013392-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR