2-Hydroxy-3-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

83430-500MG 83430-1G 83430-5G
77337-664EA 434.66 USD
77337-664 77358-912 77382-860
2-Hydroxy-3-(methylthio)benzaldehyde ≥97%
2-Hydroxy-3-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₂S
Storage Temperature: Ambient
MDL Number: MFCD16999042
CAS Number: 67868-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR