5-Bromo-1,6-dimethylpyridin-2(1H)-one ≥95%

Supplier: AOBChem USA

Warning

83394-500MG 83394-1G 83394-5G 83394-25G
77334-746EA 250.29 USD
77334-746 77358-910 77370-778 77391-388
5-Bromo-1,6-dimethylpyridin-2(1H)-one ≥95%
5-Bromo-1,6-dimethylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₇H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD22688541
CAS Number: 889865-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR