Methyl 6-bromo-5-methylbenzo[b]thiophene-2-carboxylate ≥95%

Supplier: AOBChem USA

Warning

83379-500MG 83379-1G 83379-5G 83379-25G
77334-742EA 139.05 USD
77334-742 77344-684 77370-774 77391-384
Methyl 6-bromo-5-methylbenzo[b]thiophene-2-carboxylate ≥95%
Methyl 6-bromo-5-methylbenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉BrO₂S
MW: 285.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044556
CAS Number: 1029720-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR