2,2-Dichloro-1-(3-fluoro-4-methylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

83284-500MG 83284-1G 83284-5G
77334-740EA 342.99 USD
77334-740 77358-908 77382-858
2,2-Dichloro-1-(3-fluoro-4-methylphenyl)ethanone ≥97%
2,2-Dichloro-1-(3-fluoro-4-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₇Cl₂FO
MW: 221.056 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28408397
CAS Number: 1935424-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR