(4-Bromo-2-chloro-6-fluorophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-chloro-6-fluorobenzyl alcohol, (4-Bromo-2-chloro-6-fluorophenyl)methanol

82991-500MG 82991-1G 82991-5G 82991-25G
77334-738EA 267.8 USD
77334-738 77358-906 77370-770 77391-382
(4-Bromo-2-chloro-6-fluorophenyl)methanol ≥95%
(4-Bromo-2-chloro-6-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
MW: 239.469 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25460197
CAS Number: 1823343-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR