(4-Bromo-3-chloro-2-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

82983-500MG 82983-1G 82983-5G 82983-25G
77334-734EA 90.64 USD
77334-734 77342-408 77370-768 77382-852
(4-Bromo-3-chloro-2-methylphenyl)(methyl)sulfane ≥97%
(4-Bromo-3-chloro-2-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈BrClS
MW: 251.571 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22489456
CAS Number: 127049-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR