N-(4-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pivalamide ≥97%
Supplier: AOBChem USA
|
Warning
|
82915-500MG
82915-1G
82915-25G
82915-5G
77334-732EA
300.76
USD
77334-732
77358-902
77391-380
77382-846
N-(4-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pivalamide ≥97%
N-(4-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pivalamide
Precaución: for Research Use Only
Formula:
C₁₈H₂₈BNO₃ MW: 317.231 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32661429 CAS Number: 2246631-55-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...