N-(4-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanesulfonamide ≥97%

Supplier: AOBChem USA

Warning

82914-500MG 82914-1G 82914-5G
77337-802EA 444.96 USD
77337-802 77358-900 77382-844
N-(4-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanesulfonamide ≥97%
N-(4-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanesulfonamide

Precaución: for Research Use Only

Formula: C₁₄H₂₂BNO₄S
Storage Temperature: Ambient
MDL Number: MFCD22494198
CAS Number: 710348-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR