5-Bromo-2-(ethylthio)-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

82886-500MG 82886-1G 82886-5G 82886-25G
77334-730EA 121.54 USD
77334-730 77343-884 77370-760 77382-840
5-Bromo-2-(ethylthio)-4-methylpyridine ≥97%
5-Bromo-2-(ethylthio)-4-methylpyridine

Precaución: for Research Use Only

Formula: C₈H₁₀BrNS
MW: 232.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26142806
CAS Number: 1429341-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR