5-Bromo-4-methyl-2-propoxypyridine ≥97%

Supplier: AOBChem USA

Warning

82883-500MG 82883-1G 82883-5G 82883-25G
77334-726EA 109.18 USD
77334-726 77343-228 77370-756 77382-838
5-Bromo-4-methyl-2-propoxypyridine ≥97%
5-Bromo-4-methyl-2-propoxypyridine

Precaución: for Research Use Only

Formula: C₉H₁₂BrNO
MW: 230.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29112978
CAS Number: 1289156-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR