1-(2,3-Difluoropyridin-4-yl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

82875-500MG 82875-1G 82875-5G 82875-25G
77334-724EA 227.63 USD
77334-724 77358-896 77370-754 77391-376
1-(2,3-Difluoropyridin-4-yl)-2,2-difluoroethanone ≥95%
1-(2,3-Difluoropyridin-4-yl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₇H₃F₄NO
MW: 193.098 g/mol
Storage Temperature: Ambient
CAS Number: 2353929-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR