(4-Methoxy-3,5-dimethylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

82837-500MG 82837-1G 82837-5G 82837-25G
77334-720EA 243.08 USD
77334-720 77358-894 77370-750 77391-372
(4-Methoxy-3,5-dimethylphenyl)(methyl)sulfane ≥97%
(4-Methoxy-3,5-dimethylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₄OS
MW: 182.283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617759
CAS Number: 90925-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR