2-Chloro-3-methoxy-5-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

82814-500MG 82814-1G 82814-5G 82814-25G
77334-716EA 115.36 USD
77334-716 77343-404 77370-744 77382-834
2-Chloro-3-methoxy-5-(methylthio)pyridine ≥97%
2-Chloro-3-methoxy-5-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₇H₈ClNOS
Storage Temperature: Ambient
MDL Number: MFCD34769362
CAS Number: 2901066-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR