Ethyl 2-(2-(benzyloxy)phenyl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

82794-500MG 82794-1G 82794-25G 82794-5G
77334-714EA 307.97 USD
77334-714 77358-890 77391-368 77382-832
Ethyl 2-(2-(benzyloxy)phenyl)-2-oxoacetate ≥97%
Ethyl 2-(2-(benzyloxy)phenyl)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₁₇H₁₆O₄
MW: 284.307 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11555547
CAS Number: 1256466-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR