Ethyl 2-(2,6-difluoro-3-methylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82759-1G 82759-5G 82759-25G
77342-052EA 111.24 USD
77342-052 77370-740 77382-828
Ethyl 2-(2,6-difluoro-3-methylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(2,6-difluoro-3-methylphenyl)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₁₁H₁₀F₂O₃
MW: 228.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769357
CAS Number: 1891551-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR