Ethyl 2-(6-bromo-2-fluoro-3-methoxyphenyl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

82736-1G 82736-5G 82736-25G
77342-050EA 111.24 USD
77342-050 77370-738 77382-826
Ethyl 2-(6-bromo-2-fluoro-3-methoxyphenyl)-2-oxoacetate ≥97%
Ethyl 2-(6-bromo-2-fluoro-3-methoxyphenyl)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrFO₄
MW: 305.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769346
CAS Number: 1892190-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR