Ethyl 2-(3-bromo-2-chloro-6-fluorophenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82724-500MG 82724-1G 82724-5G 82724-25G
77334-712EA 141.11 USD
77334-712 77344-942 77370-736 77391-366
Ethyl 2-(3-bromo-2-chloro-6-fluorophenyl)-2-oxoacetate ≥95%
Ethyl 2-(3-bromo-2-chloro-6-fluorophenyl)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₁₀H₇BrClFO₃
MW: 309.516 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769337
CAS Number: 1897041-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR