Ethyl 2-(3-bromo-6-chloro-2-fluorophenyl)-2-oxoacetate ≥95%
Supplier: AOBChem USA
|
Warning
|
82708-500MG
82708-1G
82708-5G
82708-25G
77334-710EA
263.68
USD
77334-710
77358-888
77370-734
77391-364
Ethyl 2-(3-bromo-6-chloro-2-fluorophenyl)-2-oxoacetate ≥95%
Ethyl 2-(3-bromo-6-chloro-2-fluorophenyl)-2-oxoacetate
Precaución: for Research Use Only
Formula:
C₁₀H₇BrClFO₃ Storage Temperature: Ambient |
MDL Number:
MFCD34769329 CAS Number: 1898081-51-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...