Ethyl 2-(3-chloro-2,4-difluorophenyl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

82695-500MG 82695-1G 82695-5G 82695-25G
77334-708EA 86.52 USD
77334-708 77342-276 77370-732 77382-824
Ethyl 2-(3-chloro-2,4-difluorophenyl)-2-oxoacetate ≥97%
Ethyl 2-(3-chloro-2,4-difluorophenyl)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₁₀H₇ClF₂O₃
MW: 248.611 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769324
CAS Number: 157373-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR