Ethyl 2-(2-fluoro-5-methylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82670-500MG 82670-1G 82670-5G 82670-25G
77334-706EA 252.35 USD
77334-706 77358-886 77370-730 77391-362
Ethyl 2-(2-fluoro-5-methylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(2-fluoro-5-methylphenyl)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₁₁H₁₁FO₃
MW: 210.202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26681875
CAS Number: 1510868-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR