Ethyl 2-(2-chloro-6-fluorophenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82637-1G 82637-5G 82637-25G
77342-048EA 111.24 USD
77342-048 77370-728 77382-822
Ethyl 2-(2-chloro-6-fluorophenyl)-2-oxoacetate ≥95%
Ethyl 2-(2-chloro-6-fluorophenyl)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₁₀H₈ClFO₃
MW: 230.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26913039
CAS Number: 132116-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR