Ethyl 2-(4-chloro-2-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82540-500MG 82540-1G 82540-5G 82540-25G
77334-700EA 139.05 USD
77334-700 77344-682 77370-724 77391-360
Ethyl 2-(4-chloro-2-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(4-chloro-2-(trifluoromethyl)phenyl)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₁₁H₈ClF₃O₃
MW: 280.628 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22688401
CAS Number: 1417508-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR