Ethyl 2-(4-fluoro-3,5-dimethylphenyl)-2-oxoacetate ≥97%
Supplier: AOBChem USA
|
Warning
|
82520-500MG
82520-1G
82520-5G
82520-25G
77334-698EA
150.38
USD
77334-698
77345-408
77370-722
77391-358
Ethyl 2-(4-fluoro-3,5-dimethylphenyl)-2-oxoacetate ≥97%
Ethyl 2-(4-fluoro-3,5-dimethylphenyl)-2-oxoacetate
Precaución: for Research Use Only
Formula:
C₁₂H₁₃FO₃ MW: 224.228 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11555636 CAS Number: 1256482-47-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...