Ethyl 2-(3-ethylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82516-500MG 82516-1G 82516-5G 82516-25G
77334-696EA 139.05 USD
77334-696 77344-732 77370-720 77391-356
Ethyl 2-(3-ethylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(3-ethylphenyl)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₃
MW: 206.238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11113524
CAS Number: 1373519-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR