Ethyl 2-(2,3-difluorophenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82510-500MG 82510-1G 82510-5G 82510-25G
77334-694EA 244.11 USD
77334-694 77358-882 77370-718 77391-354
Ethyl 2-(2,3-difluorophenyl)-2-oxoacetate ≥95%
Ethyl 2-(2,3-difluorophenyl)-2-oxoacetate

Precaución: for Research Use Only

Formula: C₁₀H₈F₂O₃
MW: 214.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26681876
CAS Number: 1192312-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR